Learn More
RCI Conductivity Indicators Products for Science Education
Ideal for comparing various solutions across a wide range of conductivity levels.
Manufacturer:  Exper Tech RCIJR
 View more versions of this product
Catalog No. S63198
Add to cart
Requires : 9V transistor battery
Description
Description
Simply immerse the probes in the solution and record the display lamp lit. The higher the numbered display lamp lit, the greater the solution conductivity.
  • Unique, multiple lamp conductivity display determines the relative conductivity of solutions
  • Choose from the 10-level RCI Junior or the advanced, 20-level RCI-DX conductivity indicator
Specifications
Specifications
RCI-JR (10-level RCI Junior)
Ideal for comparing various solutions across a wide range of conductivity levels
9V
LED
Conductivity