Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RCL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101529
Description
Antibody detects endogenous levels of total RCL.
RCL was identified on the basis of its stimulation by c-Myc protein. The latter is a transcription factor that participates in the regulation of cell proliferation, differentiation, and apoptosis. The exact function of this gene is not known but studies in rat suggest a role in cellular proliferation and c-Myc-mediated transformation.
Specifications
RCL | |
Polyclonal | |
Unconjugated | |
DNPH1 | |
2'-deoxynucleoside 5'-phosphate N-hydrolase 1; 2'-deoxynucleoside 5'-phosphate N-hydrolase 1 {ECO:0000255; C6orf108; C76683; c-Myc-responsive protein RCL; c-Myc-responsive protein Rcl {ECO:0000255; Deoxyribonucleoside 5'-monophosphate N-glycosidase; dJ330M21.3; DNPH1; HAMAP-Rule:MF_03036}; Nucleoside 2-deoxyribosyltransferase domain containing protein RGD620382; putative c-Myc-responsive; Rcl; RGD620382; RP3-330M21.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10591 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43598 | |
DNPH1 | |
A synthesized peptide derived from human DNPH1(Accession O43598), corresponding to amino acid residues R9-A59. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction