Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCOR2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

$436.00

Specifications

Antigen RCOR2
Applications Western Blot, Immunohistochemistry - Free Floating
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP174099
View Documents
Novus Biologicals
NBP174099
100 μL
Each for $436.00
Add to cart
 
NBP17409920
View Documents
Novus Biologicals
NBP17409920UL
20 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

RCOR2 Polyclonal specifically detects RCOR2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.
Specifications

Specifications

RCOR2
Polyclonal
Rabbit
CoREST2, REST corepressor 2
RCOR2
IgG
Affinity Purified
53 kDa
Western Blot, Immunohistochemistry - Free Floating
Unconjugated
RUO
283248
Synthetic peptides corresponding to the C terminal of Rcor2. Immunizing peptide sequence PPEANTKFNSRWTTDEQLLAVQAIRRYGKDFGAIAEVIGNKTLTQVKTFF.
Primary
Store at -20C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit