Learn More
RCOR3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301273AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
RCOR3 is a REST corepressor 3 protein located on chromosome 1.Specifications
| RCOR3 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q6PGA0, Q9P2K3 | |
| Rcor3 | |
| Between 445 and 495 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| RCOR3 | |
| FLJ10876 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 214742, 55758 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.