Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Recoverin Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549191

Catalog No. PA549191


Add to cart

Description

Description

Seems to be implicated in the pathway from retinal rod guanylate cyclase to rhodopsin. May be involved in the inhibition of the phosphorylation of rhodopsin in a calcium-dependent manner. The calcium-bound recoverin prolongs the photoresponse.
Specifications

Specifications

Recoverin
Polyclonal
PBS with 0.09% sodium azide
P34057
Recoverin, 23 kDa photoreceptor cell-specific protein, Cancer-associated retinopathy protein, Protein CAR, Rcvrn, Rcv1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
19674
Western Blot
Unconjugated
Rcvrn
Liquid
RCVRN
KLH conjugated synthetic peptide between 39-67 amino acids from the N-terminal region of mouse Rcvrn.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit