Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RD Plastics Clear Drawstring Bag
SDP

Catalog No. p-4447859
Click to view available options
Product Type:
Biohazard Bag
Drawstring Bag

Clear drawstring bags allow for easy content identification and keep contents secure. Available unprinted or with histology biohazard markings.

  • Clear LDPE transport bags with nylon drawstring closure
  • Allows for easy content identification and holds contents securely
  • Unmarked or printed with “HISTOLOGY TISSUE” and orange and black biohazard symbol

Specifications

Autoclavable Not Autoclavable
Capacity (English) 7 lb.
Closure Type Drawstring
Color Clear
For Use With (Application) For easy content identification and keeps contents secure
Material LDPE
No. per Case 500/Cs.
Thickness (English) 2 mil
Width (Metric) 38.1 cm
Product Type Drawstring Bag
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.