Learn More
RD Plastics LABTITE™ II Transport Bags
SDP
With front pouch for requisition slip; separate back pouch for specimen
Manufacturer:  RD Plastics G238
 View more versions of this product
Catalog No. 01-813-31
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Pull-away release liner and press to close — no glove-to-adhesive contact
  • Easy to open with specifically designed polymer film that allows bag to tear at scoreline without the use of knives or sharp instruments
  • Measures 6 × 10 in. (15.2 × 25.4cm)
  • Available with or without absorbent pad
Specifications
Specifications
Liquid-Tight Adhesive
10 x 6 in. (25.4 x 15.2cm)
25.4 cm
15.2 cm
With absorbent pad
10 in.
6 in.