Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDH11 Mouse anti-Human, Clone: OTI1B4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501053

Catalog No. 50-167-2196


Add to cart

Description

Description

RHD11, a member of the short-chain dehydrogenase/reductase superfamily of oxidoreductases, is expressed at high levels in prostate epithelium, and its expression is regulated by androgens.
Specifications

Specifications

RDH11
Monoclonal
0.99 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8TC12
ARSDR1, CGI82, HCBP12, MDT1, PSDR1, RALR1, RDJCSS, SCALD, SDR7C1
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51109
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1B4
Unconjugated
RDH11
Liquid
RDH11
Full length human recombit protein of human RDH11 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit