Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RDH11 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA14231

Catalog No. PA14231


Only null left
Add to Cart

Description

Description

PA1-4231 detects the PSDR1 protein in human samples. PA1- 4231 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~28 kDa protein representing PSDR1 in androgen-stimulated metastatic adenocarcinoma LNCaP.FGC cells. PA1- 4231 immunogen is a mixture of synthetic peptides corresponding to the N- and C-terminal residues M(1) V E L M F P(7) and L(312) L G L P I D(318) of human PSDR1.

RHD11, a member of the short-chain dehydrogenase/reductase superfamily of oxidoreductases, is expressed at high levels in prostate epithelium, and its expression is regulated by androgens.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RDH11
Polyclonal
Unconjugated
Rdh11
2610319N22Rik; AI428145; all-trans and 9-cis; androgen-regulated short-chain dehydrogenase/reductase 1; Arsdr1; AU045252; C85936; cell line MC/9.IL4 derived transcript 1; cell line MC/9.IL4-derived protein 1; CGI82; CGI-82; HCBP12; HCV core-binding protein HCBP12; M42C60; Mdt1; prostate short-chain dehydrogenase reductase 1; prostate short-chain dehydrogenase/reductase 1; Psdr1; ralR1; Rdh11; RDJCSS; Retinal reductase 1; retinol dehydrogenase 11; retinol dehydrogenase 11 (all-trans/9-cis/11-cis); SCALD; SDR7C1; Short chain dehydrogenase/reductase family 7C member 1; short chain dehydrogenase/reductase family 7C, member 1; short-chain aldehyde dehydrogenase; short-chain dehydrogenase/reductase 1; Ube-1c; UBE-1c1; ubiquitously expressing
Rabbit
Antigen affinity chromatography
RUO
51109
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8TC12
Rdh11
Synthetic peptides corresponding to residues M(1) V E L M F P(7) and L(312) L G L P I D(318) of human PSDR1.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.