Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDH11 Mouse anti-Human, Clone: OTI1B4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525073

Catalog No. PIMA525073


Add to cart

Description

Description

PSDR1 is a novel prostate short-chain dehydrogenase/reductase that exhibits increased expression on exposure to androgens in the LNCaP prostate cancer cell line. PSDR1 is expressed in normal and neoplastic prostate epithelium and is implicated in the pathogenesis of prostate carcinoma.
Specifications

Specifications

RDH11
Monoclonal
0.99 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8TC12
Retinol dehydrogenase 11, Retinal reductase 1, RalR1, Prostate short-chain dehydrogenase/reductase 1, Androgen-regulated short-chain dehydrogenase/red
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51109
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1B4
Unconjugated
RDH11
Liquid
RDH11
Full length human recombit protein of RDH11 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit