Learn More
Aviva Systems Biology Corp RDH12 Antibody - middle region (ARP52973_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52973_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RDH12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RDH12. Alternate names: FLJ30273, LCA3, LCA13, SDR7C2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60884