Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RDH12 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15801720UL
Description
RDH12 Polyclonal specifically detects RDH12 in Human samples. It is validated for Western Blot.Specifications
RDH12 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
Q96NR8 | |
RDH12 | |
Synthetic peptides corresponding to RDH12(retinol dehydrogenase 12 (all-trans/9-cis/11-cis)) The peptide sequence was selected from the middle region of RDH12. Peptide sequence AKRLQGTGVTTYAVHPGVVRSELVRHSSLLCLLWRLFSPFVKTAREGAQT. | |
20 μL | |
Vision | |
145226 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
All-trans and 9-cis retinol dehydrogenase, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.100, FLJ30273, LCA13retinol dehydrogenase 12, all-trans and 9-cis, LCA3, retinol dehydrogenase 12, retinol dehydrogenase 12 (all-trans and 9-cis), retinol dehydrogenase 12 (all-trans/9-cis/11-cis), SDR7C2, short chain dehydrogenase/reductase family 7C, member 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title