Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDH14 Mouse anti-Human, Rat, Clone: OTI1A7, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502478

Catalog No. 50-167-3531


Add to cart

Description

Description

Exhibits an oxidoreductive catalytic activity towards retinoids. Most efficient as an NADPH-dependent retinal reductase. Displays high activity toward 9-cis and all-trans-retinol. No steroid dehydrogenase activity detected.
Specifications

Specifications

RDH14
Monoclonal
1.2 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9HBH5
PAN2, SDR7C4
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
500629, 57665
Flow Cytometry, Western Blot
OTI1A7
Unconjugated
RDH14
Liquid
Rdh14
Full length human recombit protein of human RDH14 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit