Learn More
RDH14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 162831AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0851
Add to cart
Description
Description
Exhibits an oxidoreductive catalytic activity towards retinoids. Most efficient as an NADPH-dependent retinal reductase. Displays high activity toward 9-cis and all-trans-retinol. No steroid dehydrogenase activity detected.
Specifications
Specifications
RDH14
Polyclonal
Unconjugated
RDH14
Alcohol dehydrogenase PAN2, PAN2, RDH14, Retinol dehydrogenase 14, SDR7C4
Rabbit
Antigen Affinity Chromatography
RUO
105014, 500629, 57665
-20°C
Liquid
Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9ERI6, Q9HBH5
Rdh14
RDH14 Fusion Protein Ag9386
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG