Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Reaction Flasks - 50 mL

Heidolph™ Reaction Flasks - 50 mL for use with the Parallel Reaction Station

Manufacturer:  Heidolph™ 015880640

Catalog No. 05-403-956


Add to cart

Description

Description

  • 50 mL Reaction Flasks
  • For use with Parallel Reaction Station
Specifications

Specifications

Parallel Reaction Station
Glass
Round Bottom
Reaction Vessel
6/Pk.
50 mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit