Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Reaction Tube Flask

Flask features an outer joint and 2 mm Teflon™ stopcock.

Manufacturer:  Synthware™ F894200S

 View more versions of this product

Catalog No. 31-502-391


Add to cart

Description

Description

  • Flask features a 2 mm Teflon™ stopcock
Specifications

Specifications

Teflon™
Borosilicate Glass 3.3
Reaction Tube
Autoclavable
No
Non-sterile
Round
2 mm
200 mL
Sample Storage
24/40
No
Beaded
A
Stopper
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit