Ready America™ Fastener Kits for Countertop Equipment

Used for countertop and desktop equipment

Manufacturer: READY AMERICA RDT44B1

 View more versions of this product

Catalog No. 11-900-013

  Check Availability
Add to Cart

    Specifications

    Product Type Equipment Fastener
    Color Black
    Quantity Each
    Includes Four 2 x 2 in. buckles with 3M VHB adhesive, two 1 x 5 in. locking straps and two surface prep pads, instructions
    For Use With (Equipment) Countertop and desktop equipment that weigh less than 50 lb.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit