Learn More
Ready America™ HPLC Stack Fastener Kits
Secures HPLC stacks
Manufacturer:  READY AMERICA RDHP8G1
 View more versions of this product
Catalog No. 11-900-008
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Gray
Two 2 x 2 in. countertop buckles with adhesive, sixteen 1 x 2 in. equipment buckles with adhesive, two 12 in. coated locking straps, 16 prep pads
Each
Secure HPLC with base buckles to countertop and equipment buckles to 2 stacked components on both sides
HPLC 8 stack fastener kit