Ready America™ HPLC Stack Fastener Kits

Secures HPLC stacks

Manufacturer: READY AMERICA RDHP2G1

 View more versions of this product

Catalog No. 11-900-002

  Check Availability
Add to Cart

    Specifications

    Product Type HPLC 2 stack fastener kit
    Color Gray
    Quantity Each
    Includes Two 2 x 2 in. countertop buckles with adhesive, four 1 x 2 in. equipment buckles with adhesive, two 12 in. coated locking straps, 4 prep pads
    For Use With (Application) Secure HPLC with base buckles to countertop and equipment buckles to 2 stacked components on both sides

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit