Ready America™ Wall-Mount Safety Straps for Large Equipment

Secures kitchen-size freezers, refrigerators, incubators, centrifuges and similar type items

Manufacturer: READY AMERICA RW2X48W21

 View more versions of this product

Catalog No. 11-900-034

  Check Availability
Add to Cart

    Specifications

    Product Type Equipment Wall Strap™
    Color White
    Quantity Each
    Includes Two 2 x 48 in. straps with industrial hook and loop with side-release buckle, two 0.25 in. Wood stud screws, two 0.25 in. Metal stud togglers with instructions, two 0.25 in. Metal stud screws, two 0.25 x 1 in. washers, two surface prep pads instructions
    For Use With (Equipment) Secures kitchen size refrigerators, freezers and similar sized items to studs in walls, with easy disconnect

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit