Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ready-Made LB Agar Powder, Thermo Scientific Chemicals
For propagation of bacteria
Supplier: Thermo Scientific J75851A1
Description
pH(3.5%, H2O): 7.0 ± 0.2Contents:
- Casein Peptone: 10gm/L
- Yeast Extract: 5gm/L
- Sodium Chloride: 5gm/L
- Agar: 15gm/L
Specifications
Powder | |
Propagation of Bacteria | |
1 kg |
Agar | |
Plastic Bottle |
Safety and Handling
Recommended Storage : Ambient temperatures
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction