Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Ready-Made LB Agar Powder

For propagation of bacteria

$156.30

Specifications

Packaging Plastic Bottle
For Use With (Application) Propagation of bacteria
Format Free Flowing Powder
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ75851A1 View Documents Thermo Scientific™
J75851A1
1kg Each for $156.30
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

pH(3.5%, H2O): 7.0 ± 0.2
Contents:
  • Casein Peptone: 10gm/L
  • Yeast Extract: 5gm/L
  • Sodium Chloride: 5gm/L
  • Agar: 15gm/L
Specifications

Specifications

Plastic Bottle
Propagation of bacteria
Free Flowing Powder
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit