Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent 2 (REAG-512), Acetate Buffer, Reagents

Catalog No. p-200021781
Click to view available options
Quantity:
1 L
10 L

Reagent 2 (REAG-512), Acetate Buffer, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Reagent 2 (REAG-512), Acetate Buffer
CAS 7732-18-5, 631-61-8, 1336-21-6
Boiling Point 100°C
Color Colorless
For Use With (Equipment) Water Analyzer
Physical Form Liquid
Quantity 10 L
Solution Type Buffer
Packaging HDPE Winpak
Specific Gravity 1.0116
Sterility Non-sterile
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.