Learn More
Andwin ScientificSupplier Diversity Partner REAGENT BOTTLES/VIP-5 DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner 87001816
These reagent bottles with attachments can be used with the Tissue Tek VIP 5 Vaccuum Infiltration Tissue Processors S7643-1 and S7643-2.
Catalog No. 50-112-6000