Learn More
Thermo Scientific™ Reagent Grade Deionized Water Available on GSA/VA Contract for Federal Government customers only.
Instantly destroy bacteria with Thermo Scientific™ Reagent Grade Deionized Water.
 View more versions of this product
Catalog No. 23-751628
  Check Availability
Add to cart
Disclaimers:
Not intended for infusion, injection or irrigation.
Specifications
Description
Instantly destroy bacteria with Thermo Scientific™ Reagent Grade Deionized Water, ozonated to prevent/inhibit microbial growth.
  • For use in flame photometry, blood gas humidifying chambers, electrode maintenance and instrument cleaning
  • Classified as Reagent Grade Water by NCCLS standards
Note:
Because of the many ways customers use this product, Thermo Fisher Scientific cannot provide an open container stability claim. Customers may establish their own open container stability claim by following CLSI guidelines.
Description & Specifications
Specifications
5 gal.