Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Reagent Refills
SDPSCI_ED

Testing supplies for the discerning scientist

$10.55 - $182.57

Specifications

Storage Requirements 25°C
View More Specs

Products 15
Catalog Number Mfr. No. Type Shelf Life Volume (English) Price Quantity & Availability  
Catalog Number Mfr. No. Type Shelf Life Volume (English) Price Quantity & Availability  
S72311
SDPSCI_ED
View Documents
LaMotte™
4498L
Chlorine Reagent 1 18 Months 16.9 oz.
Each for $65.75
Only null left
Add to Cart
 
S72312
SDPSCI_ED
Accessories View Documents
LaMotte™
4500L
Chlorine Reagent 3 2 Years 16.9 oz.
Each for $34.78
Only null left
Add to Cart
 
S72318
SDPSCI_ED
View Documents
LaMotte™
4633H
Sulfide 1 2 Years 2.02 oz.
Each for $19.56
Only null left
Add to Cart
 
S72319
SDPSCI_ED
View Documents
LaMotte™
4634H
Sulfide 2 3 Years 2.02 oz.
Each for $19.56
Only null left
Add to Cart
 
S72322
SDPSCI_ED
View Documents
LaMotte™
V4797G
Ammonia Reagent 1 3 Years 1.01 oz.
Each for $16.57
Only null left
Add to Cart
 
S72310
SDPSCI_ED
View Documents
LaMotte™
4493DRH
Alkalinity Reagent B 2 Years 2.02 oz.
Each for $13.04
Only null left
Add to Cart
 
S72393
SDPSCI_ED
View Documents
LaMotte™
V6279C
Nitrogen Reducing Reagent 1 Year 0.01 lb.
Each for $17.94
Only null left
Add to Cart
 
S72384
SDPSCI_ED
View Documents
LaMotte™
4498E
Chlorine Reagent 1 18 Months 0.5 oz.
Each for $10.55
Only null left
Add to Cart
 
S72386
SDPSCI_ED
View Documents
LaMotte™
6807J
DPD 1 Powder 18 Months 0.22 lb.
Each for $60.86
Only null left
Add to Cart
 
S72394
SDPSCI_ED
View Documents
LaMotte™
V6278H
Mixed Acid 3 Years 2.02 oz.
Each for $19.83
Only null left
Add to Cart
 
S72321
SDPSCI_ED
View Documents
LaMotte™
7044AJ
Chlorine Reagent Refill 3 Years 16.9 oz.
Each for $49.17
Only null left
Add to Cart
 
S72339
SDPSCI_ED
View Documents
LaMotte™
4635H
Sulfide Test 3 2 Years 2.02 oz.
Each for $20.65
Only null left
Add to Cart
 
S72340
SDPSCI_ED
View Documents
LaMotte™
4636H
Sulfide Test 4 3 Years 2.02 oz.
Each for $17.66
Only null left
Add to Cart
 
S72344
SDPSCI_ED
View Documents
LaMotte™
6410L
Ferroin Indicator 2 Years 16.9 oz.
Each for $50.26
Only null left
Add to Cart
 
S72345
SDPSCI_ED
View Documents
LaMotte™
6521M
Potassium Iodide 2 Years 32.1 oz.
Each for $182.57
Only null left
Add to Cart
 
Description

Description

Refills for sulfide tests, potassium iodide, chlorine, ferroine indicators and more

Specifications

Specifications

25°C
Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 Months

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.