Reagents for Roche Diagnostics cobas™ c 111 Chemistry Analyzer

Manufacturer: Roche Diagnostics 04774221190

 View more versions of this product

Catalog No. 22-045-852

  Check Availability
Add to Cart

    Specifications

    Reagents Included Pyridoxal Phosphate reagent
    Product Type Pyridoxal Phosphate reagent
    For Use With (Equipment) Cobas™ c 111 chemistry analyzer
    Quantity 800 Tests
    Sensitivity 2μL
    Shelf Life 1 month
    Test Time 10.5 min.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit