Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Rearview Mirror Hanging Tags

Catalog Number 19104373
Click to view available options
Item Description:
001-100
101-200
201-300
301-400
401-500
Color:
Blue
Red

Control parking with an economical and effective solution. Brady™ Rearview Mirror Hanging Tags are a durable solution that make parking control easy.

  • Strong wording on an economical yet durable permit 
Width (English) 2.75 in.
Dimensions (L x W) 4.75 in. x 2.75 in.
Color Red
Length (English) 4.75 in.
Item Description 101-200
Material Vinyl
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit