Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ Rebreathing Small Animal Anesthesia Machine
SDP

Made of high-quality materials, supports an O2 concentrator and has a small footprint

Supplier:  SOMNI Scientific™ RM17003

Catalog No. 04-777-108

Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.


Description

Description

  • High quality limited maintenance design
  • Pressure manometer center mounted for immediate visibility
  • Toggle switch: rebreathing to nonrebreathing, circuits stay in place
  • H-frame with shelf provides support and stability
  • 3 in. wheels for maneuverability
  • Popoff valve releases at 2 cm H2O while maintaining passive volume
  • Supports oxygen concentrator
  • Small footprint, easy assembly
  • Powder coated and anodized frame for easy cleaning
Specifications

Specifications

Animal Oxygen Supply, Inhalant Anesthesia
Powder-coated and Anodized Aluminum
53.34 cm
VS-6002, VS-6001,VS-6004, VS-6006, VS-6005, VS-6008, OC-21003, RE-20101, WG-15003, DS-14005
53.34 cm
Adult & pediatric F-circuit, 2 L bag
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Veterinary Use Only