Learn More
Pepro Tech Inc Recombinant Human SCF  DFS Item
Manufacturer:  Pepro Tech Inc 30007250UG
Recombinant Human SCF is an 18.4 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF.
Catalog No. 50-162-5216