Learn More
Pepro Tech Inc Recombinant Human GITR/TNFRSF18 Fc, 20ug DFS Item
Manufacturer:  Pepro Tech Inc 31022R20UG
Recombinant Human GITR/TNFRSF18 Fc is a glycosylated, disulfide-linked homodimer of 738 a.a. residues whose monomer consists of a 136 a.a. extracellular domain fused to the 231-amino-acid length Fc portion of human IgG by two glycine residues.
Catalog No. 50-195-2526