Learn More
Biotang IncSupplier Diversity Partner Resistin Mutant Human Recombinant, A12-alpha-like 2, RSTN, XCP1, RETN1, 9.9 kDa protein containing 93 amino acid residues, produced in E.coli. has had a Cysteine residue mutated to prevent dimerization and possibly acts as an antagonist. DFS Item
Manufacturer:  Biotang IncSupplier Diversity Partner BTCYT585
Resistin Mutant Human Recombinant, A12-alpha-like 2, RSTN, XCP1, RETN1, 9.9 kDa protein containing 93 amino acid residues, produced in E.coli. has had a Cysteine residue mutated to prevent dimerization and possibly acts as an antagonist.
Catalog No. 50-150-400