Learn More
Biotang Inc Transgelin Human Recombinant, TAGLN Human, SM22, SMCC, TAGLN1, WS3-10, SM22-alpha, TAGLN, containing 221 amino acids (1-201 a.a.) and having a molecular mass of 24.8 kDa. fused to 20 amino acid His-Tag at N-terminus
Catalog No. 50150637 Shop All BIOTANG Inc Products
Encompass_Preferred
Transgelin Human Recombinant, TAGLN Human, SM22, SMCC, TAGLN1, WS3-10, SM22-alpha, TAGLN, containing 221 amino acids (1-201 a.a.) and having a molecular mass of 24.8 kDa. fused to 20 amino acid His-Tag at N-terminus
Catalog No. 50-150-637 Supplier Biotang Inc Supplier No. BTPRO851
Transgelin Human Recombinant, TAGLN Human, SM22, SMCC, TAGLN1, WS3-10, SM22-alpha, TAGLN, containing 221 amino acids (1-201 a.a.) and having a molecular mass of 24.8 kDa. fused to 20 amino acid His-Tag at N-terminus