Learn More
Pepro Tech Inc Animal-Free Recombinant Human Artemin, 500ug DFS Item
Manufacturer:  Pepro Tech Inc AF45017500UG
Recombinant Human Artemin is a 24.2 kDa, disulfide-linked homodimer formed by two identical 113 amino acid subunits.
Catalog No. 50-195-2659