Learn More
Pepro Tech Inc Animal-Free Recombinant Human Epiregulin, 25ug DFS Item
Manufacturer:  Pepro Tech Inc AF1000425UG
Recombinant Human Epiregulin is a 5.6 kDa monomeric protein, containing 50 amino residues, which corresponds to the mature secreted Epiregulin sequence.
Catalog No. 50-195-2622