Learn More
Pepro Tech Inc Recombinant Human Exodus-2 (CCL21), 20ug DFS Item
Manufacturer:  Pepro Tech Inc 30035A20UG
Recombinant Human Exodus‐2 (CCL21) is a 12.2 kDa protein containing 111 amino acid residues.
Catalog No. 50-195-2514