Learn More
Pepro Tech Inc Animal-Free Recombinant Human FGF-6, 5ug DFS Item
Manufacturer:  Pepro Tech Inc AF100305UG
Recombinant Human FGF-6 is an 18.7 kDa protein consisting of 168 amino acid residues.
Catalog No. 50-195-2630