Learn More
Pepro Tech Inc Recombinant Human TMIGD2/CD28H Fc, 1mg DFS Item
Manufacturer:  Pepro Tech Inc 310421MG
CHO cell-derived Recombinant Human TMGID2/CD28H Fc is a glycosylated, disulfide-linked homodimer of 361a.a. residues whose monomer consists of the 128a.a. extracellular domain fused to the 231a.a. length Fc portion of human IgG1 by two glycine residues.
Catalog No. 50-195-2531