Learn More
Pepro Tech Inc Animal-Free Recombinant Murine LIGHT, 20ug DFS Item
Manufacturer:  Pepro Tech Inc AF3151220UG
Recombinant Murine LIGHT is a 20.1 kDa protein containing 183 amino acid residues, comprising the TNF-homologous region of the LIGHT extracellular domain.
Catalog No. 50-195-2651