Learn More
Pepro Tech Inc Recombinant Murine Prokineticin-2, 5ug DFS Item
Manufacturer:  Pepro Tech Inc 315385UG
Recombinant Murine Prokineticin-2 is an 11.6 kDa protein consisting of 103 amino acid residues, including ten cysteine residues that can potentially form five pairs of intra-molecular disulfide bonds.
Catalog No. 50-195-2552