Learn More
Pepro Tech Inc Recombinant Murine TARC (CCL17), 100ug DFS Item
Manufacturer:  Pepro Tech Inc 25043100UG
Recombinant Murine TARC (CCL17) is an 7.9 kDa protein containing 70 amino acid residues, including the four conserved cysteine residues present in CC chemokines.
Catalog No. 50-195-2506