Learn More
Pepro Tech Inc Recombinant Murine TARC (CCL17), 20ug DFS Item
Manufacturer:  Pepro Tech Inc 2504320UG
Recombinant Murine TARC (CCL17) is an 7.9 kDa protein containing 70 amino acid residues, including the four conserved cysteine residues present in CC chemokines.
Catalog No. 50-195-2508