Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REC8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120322

This item is not returnable.
View return policy
Description
Positive test controls include: Rat testis. The target is usually found in the following locations: Chromosome, Nucleus.
REC8 Polyclonal antibody specifically detects REC8 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
REC8 | |
Polyclonal | |
Unconjugated | |
REC8 | |
cohesin Rec8p; cohesion rec8p; HR21spB; human homolog of rad21, S. pombe; kleisin-alpha; Mei8; meiosis specific sister chromatid cohesion protein; meiotic cohesion Rec8; meiotic recombination and sister chromatid cohesion phosphoprotein of the rad21p family; meiotic recombination protein REC8 homolog; meiotic recombination protein REC8-like 1; mrec; Rec8; rec8 {ECO:0000250; REC8 homolog; REC8 meiotic recombination protein; Rec8L1; REC8-like 1; Rec8p; recombination and sister chromatid cohesion protein homolog; UniProtKB:Q8C5S7} | |
Recombinant fusion protein containing a sequence corresponding to amino acids 194-561 of mouse REC8 (NP_064386.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
1.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95072, Q6AYJ4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
290227, 9985 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title