Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Cell Company Recalibration of Glass Filter 666-F4

For testing the photometric accuracy.

Supplier:  Scientific Cell Company 666F4 RECAL

Catalog No. 14-397-508


Explore available promotions
Add to Cart

Includes: DAkkS calibration certificate

Description

Description

  • Recalibration standard for spectrophotometer
  • Nominal Absorbance: 1
  • Certified at wavelengths: 440, 465, 546.1, 590, 635nm slit width
  • 1nm serial no.
  • Type: Solid
  • Individual filter
  • Light Path: Neutral Density
  • Packed in high-quality storage case
Specifications

Specifications

Recalibration of Glass Filter 666-F4
Testing the photometric accuracy
DAkkS calibration certificate
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.