Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RECK Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17974020UL

 View more versions of this product

Catalog No. NBP17974020


Add to cart

Description

Description

RECK Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

RECK
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
RECK
The immunogen for this antibody is Reck. Peptide sequence SCIVGGKRKSHGTSFTIDCNVCSCFAGNLVCSTRLCLSEHSSDDDRRTFT.
104 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Western Blot
Western Blot 1:1000
NP_057887
hRECK, membrane-anchored glycoprotein (metastasis and invasion), reversion-inducing-cysteine-rich protein with kazal motifs, ST15reversion-inducing cysteine-rich protein with Kazal motifs, suppression of tumorigenicity 15 (reversion-inducing-cysteine-rich protein withkazal motifs), suppression of tumorigenicity 5 (reversion-inducing-cysteine-rich protein withkazal motifs), Suppressor of tumorigenicity 15 protein
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
8434
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit