Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sino Biological™ CD70 Recombinant Human Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Gln 39-Pro 193) of human CD70 (NP_001243.1) fused to the Fc region of human IgG1 at the N-terminus with a polypeptide linker

Manufacturer:  Thermo Scientific™ 10780H01H50

Catalog No. 10-780-H01H50


Add to cart

Description

Description

Cluster of Differentiation 70 (CD70), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized mouse CD27 at 2µg/mL (100µL/well) can bind human CD70 with a linear range of 78-1250pg/mL
  • 2. Immobilized mouse CD27-Fc at 2µg/mL (100µL/well) can bind biotinylated human CD70 with a linear range of 0.39-12.5ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
TNF (Tumor Necrosis Factor)
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
970
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CD70 molecule
IgG1 Fc Tag
CD Proteins
Assay Standard
Lyophilized
45.5kDa
50μg
Truncated/Recombinant/Cell Surface Protein/Ligand
CD70, CD70/CD27L/TNFSF7, CD70/CD27L /TNFSF7G, TNFSF7
CD70
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.