Learn More
Pepro Tech Inc Recombinant Human IL-12 p80, 50ug DFS Item
Manufacturer:  Pepro Tech Inc 20012P80H50UG
Recombinant Human IL-12 p80 is an 80.0 kDa disulfide linked homodimer consisting of two p40 chains of IL-12.
Catalog No. 50-195-2665