Learn More
Pepro Tech Inc Recombinant Human IL-18BP Fc, 500ug DFS Item
Manufacturer:  Pepro Tech Inc 20018BP500UG
CHO cell-derived Recombinant Human IL-18BP Fc is a glycosylated, disulfide-linked homodimer of 792 a.a. residues whose monomer consists of a 163a.a. portion of secreted IL-18BP fused to the 231a.a. length Fc portion of hIgG by two glycine residues.
Catalog No. 50-195-2479