Learn More
Pepro Tech Inc Recombinant Human Nesfatin-1, 200ug DFS Item
Manufacturer:  Pepro Tech Inc 30067200UG
Recombinant Human Nesfatin-1 is a 9.7 kDa protein containing 82 amino acid residues.
Catalog No. 50-195-2666