Learn More
Pepro Tech Inc Recombinant Human Wnt-7a, 100ug DFS Item
Manufacturer:  Pepro Tech Inc 12031100UG
Recombinant Human Wnt-7a is a 35.5 kDa glycoprotein containing 318 amino acids. Due to glycosylation, Wnt-7a migrates between 40-55 kDa by SDS-PAGE gel under unreduced conditions.
Catalog No. 50-195-2663